Collects pipette tips, microcentrifuge tubes. Bel Art™ Poxy Coated Wire Framed Bag Holder is economical and convenient.
Have Questions?
Catalog Number
Color
FKS10633305
Orange
Catalog number FKS10633305
Price (SGD)
161.00
EA
Add to cart
Color:
Orange
Price (SGD)
161.00
EA
Add to cart
Collects pipette tips, microcentrifuge tubes. Bel Art™ Poxy Coated Wire Framed Bag Holder is economical and convenient.
Rectangular steel wire frame holds a polyethylene bag 22 x 28cm (8½ x 11 in.); coated in easy to clean, durable, Orange color epoxy
Snug fitting polypropylene plastic cover helps hold the bag securely in place
Reusable; can be cleaned by hand or in automated dish washers; autoclavable.
Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes
Can be used with Bench Top Biohazard Bags (F13166-0000); can also be used with Clavies Autoclavable Bags (H13182-0812)
Specifications
ColorOrange
For Use With (Equipment)For bench top biohazard bags