Nongridded Sterile Cellulose Membrane Filters
Nongridded Sterile Cellulose Membrane Filters
Actual product may vary
Fisher Catalogue
Nongridded Sterile Cellulose Membrane Filters
Ensure quality control in routine microbiological filtering. Sartorius Nongridded Sterile Cellulose Membrane Filters are single-packed membranes used for colony counting, particle testing, microscopy and sterility testing. Choose between cellulose acetate or cellulose nitrate membranes in a variety of pore sizes.
Have Questions?
Catalog NumberMaterialPore SizeDiameter (Metric)No. per Pack
FKS10632042Cellulose Acetate0.2 μm47 mm100/Pk.
Catalog number FKS10632042
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material:
Cellulose Acetate
Pore Size:
0.2 μm
Diameter (Metric):
47 mm
No. per Pack:
100/Pk.
Ensure quality control in routine microbiological filtering. Sartorius Nongridded Sterile Cellulose Membrane Filters are single-packed membranes used for colony counting, particle testing, microscopy and sterility testing. Choose between cellulose acetate or cellulose nitrate membranes in a variety of pore sizes.
Sterile, individually packaged filters for strict quality control
  • Pre-sterilized via gamma irradiation at 25kGray
  • Ready-to-use to save preparatory time
  • Avoids contamination of remaining filters in opened packs
  • Filter identification and lot number are printed on each individual envelope
  • Each membrane is checked to ensure:
    • It is not damaged in any way
    • It is positioned correctly with no slippage under the edge seal
    • It is free of particles
    • Each envelope has readable lettering
High-quality membranes for a variety of lab applications
  • Defined particle retention and outstanding, reliable recovery rates for microorganisms
  • Choice of two membrane materials:
    • Cellulose nitrate filters (mixed cellulose ester) assure effective retention combined with high flow rates and optimal colony growth
    • Cellulose acetate filters combine high flow rates and thermal stability with very low adsorption characteristics
  • Available in 7 different pore sizes: 0.2μm, 0.45μm, 0.65μm, 0.8μm, 1.2μm, 3.0μm and 8.0μm
    • 8μm filters are used as prefilters in a prefilter attachment. They retain coarse particles, while allowing microorganisms through for recovery on a bacteria-retentive filter
    • 0.2μm filters are the standard for sterile filtration and are validated by the Bacteria Challenge Test (BCT)
    • 0.45μm filters are manufactured according to ISO 7704
  • White, un-gridded filters in 47 or 50mm (1.9 or 2 in.) diameter sizes
Specifications
ColorWhite
DisposableSingle-use
Filter TypeMembrane Filter
FormatDisc
MaterialCellulose Acetate
Material (Membrane)Cellulose Acetate
Pore Size0.2 μm
SterilitySterile
For Use With (Application)For colony count; sterility testing; particle count; microscopy
HydrophobicityHydrophilic
Individually wrappedYes
No. per Pack100/Pk.
TypeMembrane Filter
Diameter (English)1.9 in.
Diameter (Metric)47 mm
Unit SizePK