Check Order Status
Azlon™ Polypropylene Volumetric Flask
Actual product may vary
Fisher Catalogue
Azlon™ Polypropylene Volumetric Flask
Manufactured from translucent polypropylene. Azlon™ Polypropylene Volumetric Flask features a clear easy to read calibration line.
Have Questions?
Catalog Number
Capacity (Metric)
Height (Metric)
Tolerance
FKS10626855
25 mL
115 mm
±0.08 mL
Catalog number
FKS10626855
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
25 mL
Height (Metric):
115 mm
Tolerance:
±0.08 mL
Manufactured from translucent polypropylene giving good durability
Clear easy to read calibration line
Calibrated to ISO 1042, class B tolerances
Supplied with polypropylene stopper
Specifications
Certifications/Compliance
ISO 1042
Closure Material
Polypropylene
Color
Translucent
Capacity (Metric)
25 mL
Material
Polypropylene
Height (Metric)
115 mm
Includes
Stopper
No. per Pack
1
Tolerance
±0.08 mL
Class
B
Closure Type
Stopper
Diameter (Metric) Outer
39 mm