Nalgene™ Autoclavable PPCO Wash Bottles
Nalgene™ Autoclavable PPCO Wash Bottles
Actual product may vary
Nalgene™ Autoclavable PPCO Wash Bottles
Nalgene™ Autoclavable PPCO Wash Bottles
Nalgene™ Autoclavable PPCO Wash Bottles
Nalgene™ Autoclavable PPCO Wash Bottles
Fisher Catalogue
Nalgene™ Autoclavable PPCO Wash Bottles
Reduce the risk of contamination when dispensing sterile water or reagents by using these autoclavable PPCO wash bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS10618101500 mL28 mm
2405-0500
also known as 2405-0500C
500 mL28 mm
FIS03-409-14A500 mL28 mm
Catalog number FKS10618101
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Closure Size:
28 mm
Reduce the risk of contamination when dispensing sterile water or reagents by using autoclavable Thermo Scientific™ Nalgene™ PPCO Wash Bottles. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. These squeeze bottles are flexible, contact-clear, and designed to dispense through the top with a closure and stem molded in one piece for reliable service. Bent stem dispensing tube retains its bend after autoclaving but bend angle can be modified by hand while warm if desired. The tight fit of the draw tube and closure prevents separation when squeezing bottle.
  • Autoclavable
  • Polypropylene screw/stem and draw tube
  • Tip can be cut back to provide increased flow
  • Sputter-free, precision stream
  • Bottle and Carton are recyclable in many communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
IncludesScrew/stem and draw tube
MaterialPPCO
No. per Case24
No. per Pack6
SterilityNon-sterile
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Height (Metric)213 mm
Product TypeWash Bottle
Height (English)8.4 in.
Closure MaterialPolypropylene
Closure Size28 mm
Diameter (Metric)72.6 mm
Closure TypeCap
Through the Closure DispensingYes
Unit SizePK