HDPE Narrow Neck Round Bottles
Actual product may vary
Fisherbrand
HDPE Narrow Neck Round Bottles
Store a wide range of liquids with these durable plastic bottles. Azlon™ HDPE Narrow Neck Round Bottles feature good general chemical resistance, and are suitable for food contact applications in accordance with EU and USA regulations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)No. per Pack
FKS10616865125 mL10/PK
FKS1060801530 mL10/PK
Catalog number FKS10616865
Price (SGD)
55.00
Pack
Capacity (Metric):
125 mL
No. per Pack:
10/PK
Price (SGD)
55.00
Pack
Store a wide range of liquids with these durable plastic bottles. AzlonTM HDPE Narrow Neck Round Bottles feature good general chemical resistance, and are suitable for food contact applications in accordance with EU and USA regulations.
  • Translucent, rigid HDPE bottles with polypropylene screw caps
  • Lightweight but very durable in use
  • Batch certificate available to download online
  • Bulk packs contain an equal number of bottles and caps, packed separately
Specifications
Capacity (Metric)125 mL
ColorTranslucent
Diameter (Metric) Inner Neck18 mm
MaterialHDPE
No. per Pack10/PK
Closure ColorTranslucent
Closure IncludedYes
MouthNarrow
ShapeRound
Height (Metric)102 mm
Closure MaterialPolypropylene
Diameter (Metric)50 mm
Closure TypeScrew Cap
Unit SizePack