Nalgene™ Polypropylene Floating Microtube Racks
Nalgene™ Polypropylene Floating Microtube Racks
Actual product may vary
Nalgene™ Polypropylene Floating Microtube Racks
Nalgene™ Polypropylene Floating Microtube Racks
Nalgene™ Polypropylene Floating Microtube Racks
Fisher Catalogue
Nalgene™ Polypropylene Floating Microtube Racks
Thaw, cool and incubate samples at elevated or reduced temperatures using these floating microtube racks, which are especially useful in genetic cloning, microbiology and immunology labs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPositionsShape
FKS106139918Round
5974-0404
also known as 5974-0404C
16Square
FIS14-809-16616Square
5974-101520Round
FIS14-809-166A20Round
FKS1048869220Round
5974-4015
also known as 5974-4015P
8Round
Catalog number FKS10613991
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Positions:
8
Shape:
Round
Thaw, cool and incubate samples at elevated or reduced temperatures with Thermo Scientific™ Nalgene™ Polypropylene Floating Microtube Racks. Especially useful in genetic cloning, microbiology and immunology labs, these racks offer excellent temperature resistance from -70° to +100°C (-94° to +212°F). Tubes are supported in the rack below the lip. Square or round racks have molded-in letters and numbers at each hole for easy sample identification.
Features:
  • Square- and round-style racks for incubating 0.5 to 2.0mL microcentrifuge tubes at elevated or reduced temperatures in water or ice baths
  • Round-style racks fit conveniently inside 400mL or 1000mL beakers
  • Square racks are designed for stable flotation in turbulent water baths — racks will float with a full load of filled tubes
  • On benchtop, rack legs prevent bottom of tubes from touching bench surface
  • Handle provides convenient transport, agitation and removal from water bath
  • Each hole has molded-in letters and numbers for easy sample identification
Note:
Racks are not autoclavable.
Nalgene reusable polypropylene(PP) products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These racks and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene products are packaged. And at end of product life, please wash and recycle your Nalgene racks if services are available in your community.
Specifications
Product TypeFloating Microtube Rack
Array8
AutoclavableNot Autoclavable
ColorWhite
HoldsMicrotubes (1 to 2 mL)
MaterialPolypropylene
Positions8
ShapeRound
For Use With (Equipment)Tube sizes: 1.0, 1.2, 1.5 and 2.0mL
No. per Case16
No. per Pack4
Diameter (Metric) Inner66 mm
Unit SizePK