Zirconium Oxide Grinding Balls
Zirconium Oxide Grinding Balls
Actual product may vary
Fisherbrand
Zirconium Oxide Grinding Balls
Have Questions?
Catalog NumberFor Use With (Equipment)Diameter (Metric)
FKS10608751Cell and Tissue Disruption with 200,400 mm2 mm
Catalog number FKS10608751
Price (SGD)
4,944.00
EA
Add to cart
For Use With (Equipment):
Cell and Tissue Disruption with 200,400 mm
Diameter (Metric):
2 mm
Price (SGD)
4,944.00
EA
Add to cart
Specifications
Product TypeGrinding Balls
Diameter (Metric)2 mm
For Use With (Equipment)Cell and Tissue Disruption with 200,400 mm
MaterialZirconium Oxide
Unit SizeEA