Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Actual product may vary
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Fisherbrand
Nalgene™ PPCO Amber Micro Packaging Vials: Sterile, Bulk Pack
Protect light sensitive reagents with these PPCO amber micro packaging vials, molded from high purity, noncytotoxic, nonpyrogenic, low-metal content polypropylene copolymer (PPCO) resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FKS106022542 mL
342805-00050.5 mL
342805-00202 mL
FIS03-390-272 mL
Catalog number FKS10602254
Price (SGD)
655.00
PK
Add to cart
Volume (Metric):
2 mL
Price (SGD)
655.00
PK
Add to cart
Protect light sensitive reagents with Thermo Scientific™ Nalgene™ PPCO Amber Micro Packaging Vials. These noncytotoxic and nonpyrogenic vials are molded from sterile high purity, low-metal content polypropylene copolymer resin and are ready to use. Vials are skirted and feature a conical bottom, allowing recovery of entire contents.
Features:
  • Vials and closures (listed separately) pressure-tested together at 7.5 PSIG
  • USP Class VI compliant; non-pyrogenic
  • Can be centrifuged at 13,000xg
  • Components provided sterile and ready to fill
  • Colored closures packaged separately
  • Sterile to 10-6 SAL
  • RNase/DNase free
  • Are leakproof† when used with Nalgene closures
Ordering Information:
Micro Packaging Vials are bulk packed without closures, 500 per inner bag, 1,000 per case. Order sterile closures separately (Cat. Nos. 342821, 342825).
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
DescriptionMicro Packaging Vials
Diameter (Metric) Inner8 mm
ColorAmber
Diameter (Metric) Outer13 mm
MaterialPPCO
SterilitySterile
Volume (Metric)2 mL
No. per Case1000
No. per Pack500
StyleSkirted vial with conical bottom
Volume (Metric) Total2 mL
Closure MaterialPPCO
Closure TypeScrew Cap (Not Included)
Height (Metric)49 mm
Quantity500 Bag, 1000 Cs.
Certifications/ComplianceISO 13485:2016
TypeVial
Closure Size11 mm
Unit SizePK