Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Actual product may vary
Fisherbrand
Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)Quantity
FKS10598741ISA SolutionCarbon Dioxide Electrode475 mL
FIS13-641-748Ionic Strength Adjustor BufferSodium ISE475 mL
FIS13-641-749Reconditioning SolutionSodium Electrode475 mL
FIS13-641-927ISA SolutionPotassium Electrode475 mL
FIS13-641-851ISA SolutionCalcium Electrode475 mL
FIS13-641-852ISA SolutionBromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode475 mL
FIS13-642-578TISAB IIFluoride Electrode1 Gal.
FIS13-641-874TISAB IIIFluoride Electrode475 mL
FIS13-641-770ISA SolutionCarbon Dioxide Electrode475 mL
FIS13-641-853Alkaline ReagentCyanide Electrode475 mL
Catalog number FKS10598741
Price (SGD)
-
Type:
ISA Solution
For Use With (Equipment):
Carbon Dioxide Electrode
Quantity:
475 mL
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.
Warranty: Out of box
Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.
All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:
  • Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
  • Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
  • Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
  • Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionISA for Carbon Dioxide Electrode
For Use With (Equipment)Carbon Dioxide Electrode
TypeISA Solution
For Use With (Application)Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
PackagingBottle
Quantity475 mL
Unit SizeEA