Orion™ ROSS Ultra™ Glass Bodied Reference Electrode
Orion™ ROSS Ultra™ Glass Bodied Reference Electrode
Actual product may vary
Fisherbrand
Orion™ ROSS Ultra™ Glass Bodied Reference Electrode
Perform precise pH determinations for routine or research work.
Have Questions?
Catalog NumberLength (Metric)
FKS10596669120 mm
Catalog number FKS10596669
Price (SGD)
1,274.00
EA
Add to cart
Length (Metric):
120 mm
Price (SGD)
1,274.00
EA
Add to cart
This Reference Half-Cell Electrode is for use with ROSS Half-Cell pH Electrode 8101BNWP or 8411BN ROSS sodium half-cell electrodes.
Thermo Scientific™ Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonymous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
24 months from date of purchase
Specifications
Description800500U ROSS Ultra Reference Electrode
JunctionCeramic
MaterialGlass Body
pH Range0 to 14
Temperature (Metric) Operating0°C to 100°C
TypeReference Electrode
ElectrodeReference half-cell glass body electrode; Pin tip connector
Filling Solution TypeROSS electrode fill solution
For Use With (Application)Precise pH determinations
Internal ReferenceROSS
Precision0.01
Length (Metric)120 mm
Diameter (Metric) Body120 mm
Diameter (Metric) Cap16 mm
Unit SizeEA