Ring With Bossehead
Ring With Bossehead
Actual product may vary
Ring With Bossehead
Ring With Bossehead
Fisherbrand
Ring With Bossehead
Manufactured from zinc alloy steel.
Have Questions?
Catalog NumberDiameter (Metric) Inner
FKS10595974150 mm
Catalog number FKS10595974
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Inner:
150 mm
  • Clamping nuts for rods up to 16mm in diameter
  • Accessory item
  • Stem length to the center of the bosshead is approximately 60mm
  • Bosshead in alloy, diecast on to the ring stem and fitted with a blue plastic headed nickel plated steel screw
Specifications
Diameter (Metric) Inner150 mm
IncludesWith Bossehead
Product TypeRing With Bossehead
Unit SizePK