Check Order Status
Basic Line Conductivity Standard, 84μS/cm, Hamilton™
Element Search
Structure Search
Fisherbrand
Basic Line Conductivity Standard, 84μS/cm, Hamilton™
Provides unprecedented stability. Hamilton™ Basic Line Conductivity Standard, 84μS/cm comes with a certificate with calibration document form DFM.
Have Questions?
Catalog Number
Quantity
FKS10595673
500 mL
Catalog number
FKS10595673
Price (SGD)
271.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
271.00
EA
Add to cart
Specifications
Packaging
500mL calpack bottle
Quantity
500 mL
Unit Size
EA
Remains stable for up to 3 years
Expiration date shown on every bottle
Bottles are permitted to stay open for a total of 60 minutes