No-Wire Test Tube Half Rack
No-Wire Test Tube Half Rack
Actual product may vary
Fisher Catalogue
No-Wire Test Tube Half Rack
Compact size polypropylene plastic racks save space in water baths, laminar flow hoods and fume hoods.
Have Questions?
Catalog NumberPositions
FKS1059505430
Catalog number FKS10595054
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Positions:
30
  • Polypropylene construction
  • Submersible
  • Works well in water baths
  • Empty racks can be stacked
  • Steam autoclavable at 121°C (250°F)
Note: Do not use with liquid nitrogen
Specifications
Product TypeTest Tube Half Rack
Array5 x 6
AutoclavableAutoclavable
ColorWhite
HoldsTubes (16 mm)
MaterialPolypropylene
Positions30
For Use With (Equipment)For 13-16mm Test Tubes
Quantity1 Each
Dimensions (L x W)12.8 x 10.5cm
Dimensions (L x W x H)12.8 x 10.5 x 7.2 cm
Length (Metric)12.8 cm
Width (Metric)10.5 cm
Height (Metric)7.2 cm
Diameter (Metric) Opening16 mm
Unit SizeEA