Vacusafe™ Comfort Aspiration system
Vacusafe™ Comfort Aspiration system
Actual product may vary
Fisherbrand
Vacusafe™ Comfort Aspiration system
Collect and contain biological liquid waste. Integra Biosciences™ Vacusafe™ Comfort Aspiration system is a compact laboratory aspiration system.
Have Questions?
Catalog NumberType
FKS10590273Vacusafe Comfort Aspiration System
Catalog number FKS10590273
Price (SGD)
10,031.00
EA
Add to cart
Type:
Vacusafe Comfort Aspiration System
Price (SGD)
10,031.00
EA
Add to cart
  • Compact
  • Level detection sensor
  • Control knob for infinitely vacuum setting (range: -300mbar to 600mbar)
  • Silent operation.
  • All parts exposed to critical liquids are fully autoclavable and can be sterilized for 20 minutes at 121°C,
  • Liquids are collected in the autoclavable waste bottle.
  • Hydrophobic filter reduces the risk of accidently aspirating liquid into the pump and pump housing.
  • Available with either a 4L polypropylene bottle or a 3L glass bottle. 
Specifications
Includes4 Liter Bottle, Level detection and lid with quick tubing couplings (PVDF)
Quantity1 Each
TypeVacusafe Comfort Aspiration System
Unit SizeEA