Check Order Status
Polypropylene Funnel with Fluted Interior
Actual product may vary
Fisherbrand
Polypropylene Funnel with Fluted Interior
Polypropylene construction. Short wide stem provides smooth, quick funneling.
Have Questions?
Catalog Number
Diameter (Metric)Top
Diameter (Metric) Stem
FKS10588901
150 mm
14 mm
Catalog number
FKS10588901
Price (SGD)
46.00
EA
Add to cart
Diameter (Metric)Top:
150 mm
Diameter (Metric) Stem:
14 mm
Price (SGD)
46.00
EA
Add to cart
Polypropylene construction
Short, wide stem
Includes internal fluting
Specifications
Diameter (Metric)Top
150 mm
Diameter (Metric) Stem
14 mm
Material
Polypropylene
Length (Metric) Overall
115 mm