Check Order Status
Epoxy Diagnostic Slides
Fisherbrand
Epoxy Diagnostic Slides
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use
Have Questions?
Change view
Catalog Number
Well Count
Color
Diameter (Metric)
FKS10588681
10
Black
6.7 mm Well
FKS10484681
1
Blue
8 mm Well
FKS10529261
12
White
5 mm Well
3 Options
Catalog number
FKS10588681
Price (SGD)
308.00
PK
Add to cart
Well Count:
10
Color:
Black
Diameter (Metric):
6.7 mm Well
Price (SGD)
308.00
PK
Add to cart
Binds and keeps liquids applied to the well area
Provides wettable wells for staining and growing cell cultures
Prevents cross-contamination
Hydrophobic coatings are very durable and withstand the strongest stains
Autoclavable
Specifications
Length (Metric)
25 mm
Material
Epoxy
Thickness (Metric)
1 mm
Type
Diagnostic Slide
Well Count
10
Width (Metric)
75 mm
Color
Black
Diameter (Metric)
6.7 mm Well