Pyrex™ Borosilicate Glass Density Bottles
Pyrex™ Borosilicate Glass Density Bottles
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Density Bottles
Manufactured from Pyrex™ borosilicate glass and complies with ISO 3507 standards, Gay-Lussac type Pyrex™ Borosilicate Glass Density Bottle is adjusted and ready for use with the exact volume engraved on the bottle. Supplied with individual 10/19-sized capillary stoppers numbered to match the bottle.
Have Questions?
Catalog NumberCapacity (Metric)Tolerance
FKS1058857050 mL±3mL
Catalog number FKS10588570
Price (SGD)
626.00
PK
Add to cart
Capacity (Metric):
50 mL
Tolerance:
±3mL
Price (SGD)
626.00
PK
Add to cart
Manufactured from Pyrex™ borosilicate glass and complies with ISO 3507 standards, Gay-Lussac type Pyrex™ Borosilicate Glass Density Bottle is adjusted and ready for use with the exact volume engraved on the bottle. Supplied with individual 10/19-sized capillary stoppers numbered to match the bottle.
  • Complies with ISO 3507, Gay-Lussac type
  • Adjusted ready for use - with the exact volume engraved on the bottle
  • With individual capillary stoppers which are numbered and matched with the bottle
  • Manufactured from Pyrex™ borosilicate glass
Specifications
Capacity (Metric)50 mL
MaterialBorosilicate Glass
QuantityEach
Stopper Number10/19
Tolerance±3mL
Certifications/ComplianceISO 3507
Closure IncludedYes
Closure TypeStopper
Unit SizePK