Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric)Top | Capacity (Metric) |
---|---|---|
FKS10586931 | 100 mm | 243 mL |
FIS10-348A | 64 mm | 68 mL |
4252-0080 | 79mm | 115 mL |
FIS10-348B | 79mm | 115 mL |
FIS10-348C | 100 mm | 243 mL |
FIS10-348D | 145 mm | 716 mL |
Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.