Whatman™ Quantitative Filter Paper: Grade 540 Circles
Whatman™ Quantitative Filter Paper: Grade 540 Circles
Actual product may vary
Fisher Catalogue
Whatman™ Quantitative Filter Paper: Grade 540 Circles
Use as a gravimetric filter during analysis of metals in acidic or alkaline solutions and in the collection of hydroxides. Whatman™ Quantitative Filter Paper: Grade 540 Circles acid-treated surface prevents tearing during filtration and allows the user to scrape precipitates from the paper.
Have Questions?
Catalog NumberDiameter (Metric)
FKS10586161125 mm
Catalog number FKS10586161
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
125 mm
Use as a gravimetric filter during analysis of metals in acidic or alkaline solutions and in the collection of hydroxides. Whatman™ Quantitative Filter Paper: Grade 540 Circles acid-treated surface prevents tearing during filtration and allows the user to scrape precipitates from the paper.
Whatman hardened ashless quantitative filter papers are acid-hardened to give high wet strength and chemical resistance with extremely low ash content. They are suitable for a wide range of critical filtration procedures.
  • High-purity, acid-treated cellulose lab filter paper
  • Mechanically strong 
  • Well suited for quantitative or gravimetric analysis of strongly acidic or alkaline samples
  • Extremely low ash content (0.006% ash maximum)
  • Nominal particle retention of 8μm
  • Medium flow rate (Herzberg filtration speed 200s)
Specifications
Airflow Rate13 s/100 mL/in2
DescriptionGrade 540 Ashless General Lab Filter Paper, 125 mm circles (100 pcs)
Filter TypeHardened Ashless
FormatDisc
MaterialCotton Linters
Particle Retention8 μm
Filtration SpeedASTM, 55 seconds; Herzberg, 200 seconds
For Use With (Application)Frequently used in the gravimetric analysis of metals in acid/alkali solutions and in collecting hydroxides after precipitation by strong alkalis.
GradeGrade 540
PorosityMedium
Quantity100/Pk.
Thickness (Metric)160 μm
TypeQuantitative Filter Paper
Diameter (English)4.92 in.
Diameter (Metric)125 mm
Unit SizePK