Round-Bottom Polystyrene Test Tubes with Cell Strainer Snap Cap, 5mL
Round-Bottom Polystyrene Test Tubes with Cell Strainer Snap Cap, 5mL
Actual product may vary
Round-Bottom Polystyrene Test Tubes with Cell Strainer Snap Cap, 5mL
Designed for flow cytometry applications. Falcon™ 12x75mm Test Tube with Cell Strainer Snap Cap offer a convenient solution to prepare laboratory samples.
Have Questions?
Catalog NumberMesh Size
FKS1058580135 μm
Catalog number FKS10585801
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Mesh Size:
35 μm
  • Widely used and referenced in laboratory protocols
  • Dual position snap-cap prevents sample loss with ease of handling
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352008 nonsterile)
The foundation for consistent research results
  • Provide reliable containment of laboratory fluid samples
  • Polystyrene tubes are best suited for procedures requiring high optical clarity
  • Dual-position snap caps, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • 1,400 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
  • Tube dimensions and volumes are approximate
  • OD: 12 mm
  • Graduations: No
Specifications
Mesh MaterialNylon
ColorClear
For Use With (Application)Flow cytometry applications
IncludesCell strainer blue dual position snap cap
MaterialPolystyrene
Product TypeTest Tube
SterilitySterile, nonpyrogenicity tested to less than 0.1 EU/ mL
For Use With (Equipment)Corning FACS System
Mesh Size35 μm
Unit SizeCS