PTFE Stir Bar Retriever
Actual product may vary
Cowie Technology
PTFE Stir Bar Retriever
Use to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Alnico V magnets at its core.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS10579271350 mm
FKS10147561150 mm
Catalog number FKS10579271
Price (SGD)
56.00
Each
Length (Metric):
350 mm
Price (SGD)
56.00
Each
Use to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Alnico V magnets at its core.
  • Inert and leak-free
  • Molded in pure, FDA-approved PTFE
  • Alnico V magnets
  • Autoclavable
Specifications
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Application)Removal of Stirrer Bars from vessels
For Use With (Equipment)PTFE Stirrer Bars
Height (Metric)10mm
Diameter (Metric)10 mm
Length (Metric)350 mm
Quantity1/Ea.
Unit SizeEach