Stainless Steel Instruments Trays
Actual product may vary
Bochem
Stainless Steel Instruments Trays
Manufactured from 18/10 stainless steel. Bochem™ Stainless Steel Instruments Trays are stackable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)
FKS10579232315 x 215 x 60 mm
FKS10106750230 x 133 x 50 mm
Catalog number FKS10579232
Price (SGD)
393.00
Each
Dimensions (L x W x H):
315 x 215 x 60 mm
Price (SGD)
393.00
Each
  • Includes lid with internal handle
  • Stackable
  • 18/10 stainless steel
Specifications
Dimensions (L x W x H)315 x 215 x 60 mm
Height (Metric)60 mm
Length (Metric)315 mm
MaterialStainless Steel
TypeInstrument Tray
Width (Metric)215 mm
IncludesLid
ShapeRectangular
Unit SizeEach