Azlon™ HDPE Translucent Graduated Square Bottles with Plug Caps
Azlon™ HDPE Translucent Graduated Square Bottles with Plug Caps
Actual product may vary
Fisherbrand
Azlon™ HDPE Translucent Graduated Square Bottles with Plug Caps
Store a wide range of liquids and solids with these graduated square bottles. Azlon™ HDPE Translucent Graduated Square Bottles with Plug Caps are suitable for food contact applications in accordance with EU and USA regulations.
Have Questions?
Catalog NumberCapacity (Metric)
FKS10574915100 mL
Catalog number FKS10574915
Price (SGD)
43.00
PK
Add to cart
Capacity (Metric):
100 mL
Price (SGD)
43.00
PK
Add to cart
Store a wide range of liquids and solids with these graduated square bottles. AzlonTM HDPE Translucent Graduated Square Bottles with Plug Caps are suitable for food contact applications in accordance with EU and USA regulations.
  • Heavy duty square bottles with plug seal and screw cap
  • Easy to grip caps have toggle for tamper evident wire attachment to neck
  • Molded graduations for approximate volume measurement
  • Space saving design
Specifications
Capacity (Metric)100 mL
Capacity (English)3.38 oz.
ColorTranslucent
Diameter (Metric) Inner Neck34 mm
For Use With (Application)For liquid or solid usage
MaterialHDPE
Length (Metric)48mm
No. per Pack10/PK
Width (Metric)45mm
Closure ColorTranslucent
Certifications/ComplianceEU; USA
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthWide
ShapeSquare
Height (Metric)88 mm
Product TypeGraduated Square Bottle
Width (English)1.77 in.
Length (English)1.89 in.
Height (English)3.46 in.
Diameter (Metric)34 mm (Inner Neck)
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizePK