Komet Stir Bars
Komet Stir Bars
Actual product may vary
Fisher Catalogue
Komet Stir Bars
Generate over three times the torque of conventional stirring bars with these komet stir bars, significantly enhancing stirrer performance and versatility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionShapeDiameter (Metric)Length (Metric)
FKS10574792Komet Glide-Ring 90Sphere, Pivot ring31 mm90 mm
FIS04-979-245Komet 90Sphere25 mm90 mm
FKS10331262Komet 90Sphere25 mm90 mm
50087924Komet 15Sphere9 mm15 mm
FIS04-979-247Komet 30Sphere13 mm30 mm
FIS04-979-249Komet Glide-Ring 90Sphere, Pivot ring31 mm90 mm
Catalog number FKS10574792
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Komet Glide-Ring 90
Shape:
Sphere, Pivot ring
Diameter (Metric):
31 mm
Length (Metric):
90 mm
Generate over three times the torque of conventional stirring bars with Thermo Scientific™ Komet Stir Bars, significantly enhancing stirrer performance and applications versatility.
Features:
  • High-energy Samarium-Cobalt (SmCo) magnetic core
  • Superior mixing action at high speeds, and with viscous media
  • Evenly coated PTFE surface prevents residue deposits
  • Sterilize with steam to 249.8°F (121°C)
  • Longlife stir bars are round with wear-resistant glide ring
Ordering Information:
  • Available in assorted sizes
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionKomet Glide-Ring 90
For Use With (Equipment)Stirrer Controller
CoatingPTFE
Diameter (Metric)31 mm
Length (Metric)90 mm
MaterialPTFE
Quantity1 Ea.
ShapeSphere, Pivot ring
TypeStir Bar
Unit SizeEA