PTFE Flat-Form Evaporating Dish
PTFE Flat-Form Evaporating Dish
Actual product may vary
Fisherbrand
PTFE Flat-Form Evaporating Dish
Inert and chemically resistant. The PTFE flat-form evaporating dish is available in a variety of sizes.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS1057239250 mL60 mm20 mm
Catalog number FKS10572392
Price (SGD)
53.00
EA
Add to cart
Capacity (Metric):
50 mL
Diameter (Metric):
60 mm
Height (Metric):
20 mm
Price (SGD)
53.00
EA
Add to cart
  • Isostatically molded in pure PTFE
  • Chemically resistant and inert
  • Super smooth internal finish
  • Usable to 280°C
Specifications
Capacity (English)1.66 fl oz.
Bottom ShapeFlat
Capacity (Metric)50 mL
MaterialPTFE
ShapeRound
TypeEvaporating Dish
Interior FinishSmooth
Temperature Range (Metric)-200°C to +280°C
Height (Metric)20 mm
Diameter (Metric)60 mm
Unit SizeEA