Check Order Status
Accessory for PIPETMANTM Complete Pipetting System- CarrouselTM Pipet Stand
Actual product may vary
Fisherbrand
Accessory for PIPETMANTM Complete Pipetting System- CarrouselTM Pipet Stand
Convenient rotating stand holds upto seven single and multichannel Gilson pipets
Have Questions?
Catalog Number
For Use With (Application)
FKS10571093
Holds 7 Pipettes
Catalog number
FKS10571093
Price (SGD)
737.00
EA
Add to cart
For Use With (Application):
Holds 7 Pipettes
Price (SGD)
737.00
EA
Add to cart
Robust, stable and space saving stand
Makes bench work look neat and tidy
Heavy base of stand prevents toppling over
Specifications
Color
White
For Use With
Pipettes
Material
Polypropylene
Type
Pipette Holder
For Use With (Application)
Holds 7 Pipettes
For Use With (Equipment)
PIPETMAN™ Complete Pipetting System
Includes
1x Carrousel pipette stand
Packaging Type
Box
Quantity
1
Dimensions (L x W x H)
205 X 205 X 405 mm
Length (English)
8.1 in
Length (Metric)
205 mm
Width (English)
8.1 in
Width (Metric)
205 mm
Height (English)
15.9 in
Height (Metric)
405 mm
Weight (English)
1.98 lb
Weight (Metric)
780 g
Unit Size
EA
Contents & Storage
1x Carrousel pipette stand