Check Order Status
Type I 18/10 Stainless Steel Beaker
Actual product may vary
Fisher Catalogue
Type I 18/10 Stainless Steel Beaker
Manufactured from 18/10 stainless steel. Bochem™ Type I 18/10 Stainless Steel Beaker include a handle and spout.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS10566951
500 mL
FKS10070921
1000 mL
2 Options
Catalog number
FKS10566951
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Manufactured from 18/10 stainless steel
Conical shape
With handle and spout
Specifications
Diameter (Metric) Outer
125 mm
Capacity (Metric)
500 mL
Material
Stainless Steel (18/10)
Height (Metric)
90 mm
Includes
Handle, Spout
Type
Beaker
Graduated
Yes
Style
Conical