1x1 image pixel for data collection
Vacuboy Hand Operator
Vacuboy Hand Operator
Actual product may vary
Fisherbrand
Vacuboy Hand Operator
Convert any vacuum source into a user-friendly aspiration system. Integra Biosciences™ Vacuboy™ Hand Operators are versatile aspiration tools for aspirating liquids from virtually all laboratory vessels.
Have Questions?
Catalog number FKS10565942
Price (SGD)
720.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Price (SGD)
720.00
EA
Add to cart
VACUBOY hand operator with:
  • 1-Channel stainless aspiration adapter 40 mm
  • 1-Channel aspiration adapter for disposable tips with ejector
  • Rubber adapter for Pasteur pipettes
VACUBOY is a versatile aspiration tool that converts any vacuum source into a user-friendly aspiration system for aspirating liquids from virtually any laboratory container at the touch of a button and in a controlled way.
  • Hand piece is connected to any available vacuum source using silicone tubing and fitted with one of the different aspiration adapters
  • The perfect match is obtained when VACUBOY is connected to a VACUSAFE aspiration system
  • The large choice of adapters allows VACUBOY to handle liquids from all tubes, dishes, bottles, T-flasks or multi-well plates, suiting their form and size
  • Adapters for aspiration with Pasteur pipettes, serological pipettes, disposable pipette tips or stainless steel needles
  • Multi-channel adapters for fast emptying of multi-well plates
  • Vacuum regulation by a touch-sensitive button for controlled and smooth aspiration of liquids
  • Ergonomic, perfectly suited for long working, productive or repetitive aspiration tasks
  • Anti-drip system prevents liquid waste leakage from the tip or pipette, reducing contamination risks when working with biohazardous liquids
  • Autoclavable and sterilizable, VACUBOY allows a maximal safety in the laboratory when working with hazardous biological liquid waste
Specifications
Includes1-Channel stainless aspiration adapter, 40mm 1-Channel aspiration adapter for disposable tips with ejector, rubber adapter for Pasteur pipettes
TypeHand operator for VACUBOY Laboratory Vacuum Pump
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link