DURAN™ HPLC Bottle, GL 45, Clear, with HPLC Screw Cap, 4 Tubing Connectors, Graduated
DURAN™ HPLC Bottle, GL 45, Clear, with HPLC Screw Cap, 4 Tubing Connectors, Graduated
Actual product may vary
Fisher Catalogue
DURAN™ HPLC Bottle, GL 45, Clear, with HPLC Screw Cap, 4 Tubing Connectors, Graduated
Pressure and vacuum resistant bottle for HPLC applications.
Have Questions?
Catalog NumberCapacity (Metric)
FKS10554493500 mL
Catalog number FKS10554493
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
The DURAN™ HPLC bottle is available as a ready-made system. This comprises a special pressure and vacuum resistant bottle and a screw cap with 4 ports. This enables 3 stations to be served at the same time from one vessel. At the same time a sterile pressure equalisation can be fitted to the fourth outlet.
  • Connection of different hose diameters (1.6 and 3.2 mm)
  • Unused ports can be sealed with silicone blanking seals
  • Optional sterile pressure equalisation sets (membrane filter 0.2 μm
  • Complete system comprising: DURAN™; pressure plus laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals
Specifications
Capacity (Metric)500 mL
DescriptionHPLC Bottle complete
For Use With (Application)Pressure and vacuum resistant bottle for HPLC applications; Safe transfer of liquid media within a closed and sterile system; Supply of solvents to HPLC equipment
MaterialBorosilicate Glass
Quantity2/Pk.
Thread Size (Metric)GL 45
AutoclavableAutoclavable
Closure LinerSilicone
Closure MaterialPolypropylene
Diameter (Metric)86 mm
Unit SizePK