S-Series Heated Ultrasonic Cleaning Bath
S-Series Heated Ultrasonic Cleaning Bath
Actual product may vary
Fisherbrand
S-Series Heated Ultrasonic Cleaning Bath
With state of the art microprocessor controlled ultrasonic cleaning and sweep technology. Fisherbrand™ S-Series Ultrasonic Cleaners offer a comprehensive range of analogue ultrasonic water baths covering the requirements for laboratory research, sonochemistry and degassing applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Dimensions (D x W x H)
FKS105517831.75 L180 x 175 x 212 mm
FKS101623722.75 L179 x 300 x 214 mm
FKS102394919.5 L278 x 365 x 264 mm
Catalog number FKS10551783
Price (SGD)
2,525.00
EA
Add to cart
Capacity (Metric):
1.75 L
Dimensions (D x W x H):
180 x 175 x 212 mm
Price (SGD)
2,525.00
EA
Add to cart
  • Long lifespan from cavitation resistant stainless steel tanks
  • LED display showing set and remaining time and pre-set and actual temperature
  • Newly developed cover reduces noise and accelerates the heating up process
  • Easy draining of cleaning liquid though the drain duct via a dial on the side
  • Operating panel arranged so that liquid cannot enter the electronics
  • Degas function key
Specifications
MaterialV2A Stainless Steel
Voltage230 VAC
Capacity (Metric)1.75 L
Dimensions (D x W x H)180 x 175 x 212 mm
Electrical Requirements230VAC, 95w
For Use With (Application)Laboratory, medical sector, dental laboratory, jewellery, watch sector, optician, workshop and service, industry
ModelElmasonic S 15 (H)
Temperature (Metric) Range30°C to 80°C
Weight (Metric)2.1 kg
Unit SizeEA