Heavy-Walled Narrow Neck Carboy Bottles, HDPE
Heavy-Walled Narrow Neck Carboy Bottles, HDPE
Actual product may vary
Fisherbrand
Heavy-Walled Narrow Neck Carboy Bottles, HDPE
Suitable for food stuffs; Autoclavable.
Have Questions?
Catalog NumberCapacity (Metric)
FKS105448515 L
Catalog number FKS10544851
Price (SGD)
235.00
PK
Add to cart
Capacity (Metric):
5 L
Price (SGD)
235.00
PK
Add to cart
  • Very resistant
  • O-ring seal cap make it leakproof
  • The cap can be wired down for added safety
  • Includes carrying handles
  • Suitable for foodstuff
Specifications
AutoclavableNot Autoclavable
Capacity (English)1.3 gal.
Capacity (Metric)5 L
Certifications/ComplianceFDA
DescriptionWith O-ring seal cap
Diameter (Metric) Inner Neck52.5 mm
Diameter (Metric) Outer170 mm
For Use With (Application)Suitable for food stuffs
Height (Metric) With Closure345 mm
IncludesClosure, Carrying Handles
MaterialHDPE
Mouth StyleNarrow
Neck StyleNarrow
No. per Pack4 Pk.
Product TypeHeavy-walled Carboy
Unit SizePK