Stainless Steel Instruments Tray
Actual product may vary
Fisherbrand
Stainless Steel Instruments Tray
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS10543204270 x 210 x 40 mm
Catalog number FKS10543204
Price (SGD)
332.00
Each
Dimensions (L x W x H):
270 x 210 x 40 mm
Price (SGD)
332.00
Each
Specifications
Dimensions (L x W)270 x 210 mm
Dimensions (L x W x H)270 x 210 x 40 mm
Height (Metric)40 mm
Length (Metric)270 mm
MaterialStainless Steel
TypeInstrument Tray
Width (Metric)210 mm
IncludesLid
Quantity1 Each
ShapeRectangular
Unit SizeEach