Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FKS10539082 | 250 mL |
FIS07-200-420 | 125 mL |
CRG8390 | 250 mL |
FIS07-200-446 | 250 mL |
CRG8393 | 500 mL |
FIS07-200-448 | 500 mL |
FKS10179200 | 500 mL |
FIS07-200-450 | 1 L |
FKS10177500 | 1 L |