MF-Millipore™ Membrane Filter, 0.8 μm, Gridded
MF-Millipore™ Membrane Filter, 0.8 μm, Gridded
Actual product may vary
Fisherbrand
MF-Millipore™ Membrane Filter, 0.8 μm, Gridded
Mixed cellulose esters (MCE) membrane, hydrophilic
Have Questions?
Catalog NumberDiameter (Metric)Color
FKS1053825247 mmWhite
Catalog number FKS10538252
Price (SGD)
882.00
PK
Add to cart
Diameter (Metric):
47 mm
Color:
White
Price (SGD)
882.00
PK
Add to cart
Mixed cellulose esters (MCE) membrane, hydrophilic
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
AutoclavableNot Autoclavable
Bubble Point≥1.0 bar, air with water
ColorWhite
Filter TypeMembrane Filter
MaterialMCE
Pore Size0.8 μm
SterilityNon-sterile
Flow Rate190 mL/min x cm2
For Use With (Application)For air monitoring, bioassays, dairy microbiology, particle monitoring, and particle removal
Gas Flow Rate16 L/min/ cm2
HydrophobicityHydrophilic
Membrane TypeGridded
No. per Case100Pack
Quantity100/Pk.
Thickness (Metric)150 μm
TypeFilter Membranes
WettabilityHydrophilic
Diameter (Metric)47 mm
Unit SizePK