Quantofix™ Copper Test Strip Kits
Quantofix™ Copper Test Strip Kits
Actual product may vary
Fisherbrand
Quantofix™ Copper Test Strip Kits
Allows for the quick and easy determination of copper in solutions. Macherey Nagel™ Quantofix™ Copper Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Have Questions?
Catalog NumberColor Indicator
FKS10536732White to Red/Violet
Catalog number FKS10536732
Price (SGD)
136.00
PK
Add to cart
Color Indicator:
White to Red/Violet
Price (SGD)
136.00
PK
Add to cart
Allows for the quick and easy determination of copper in solutions. Macherey Nagel™ Quantofix™ Copper Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
  • Ready to use
  • Rapid: provides reliable results within 20 seconds
  • Excellent for the electroplating process and water sampling
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Self life: 2.5 years, store below 30°C
Specifications
Color IndicatorWhite to Red/Violet
FormatStrip
For Use With (Application)Determination of Copper
Graduations0, 10, 30, 100, 300mg/L Cu1+/2+
Quantity100 Pack
Shelf Life2.5 Years
Storage Requirements30°C
Test Time20 sec.
TypeSemi-qualitative Test Strip
Unit SizePK