Quickfit™ Borosilicate Glass Cold Trap Head
Quickfit™ Borosilicate Glass Cold Trap Head
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Cold Trap Head
Freezes volatile materials and vapors onto the cold surface. Pyrex™ Quickfit™ Borosilicate Glass Cold Trap Head prevents materials from damaging vacuum pumps.
Have Questions?
Catalog number FKS10536465
Price (SGD)
548.00
EA
Add to cart
Price (SGD)
548.00
EA
Add to cart
Freezes volatile materials and vapors onto the cold surface. Pyrex™ Quickfit™ Borosilicate Glass Cold Trap Head prevents materials from damaging vacuum pumps.
  • Designed for use in vacuum systems
  • Prevents vapours from volatile materials from damaging pumps etc
  • Easy to use: - immerse trap into Dewar vessel containing refrigerant e.g., dry ice - volatile materials passing through system are frozen onto the cold surface
Specifications
DescriptionCold trap head
MaterialBorosilicate Glass
For Use With (Equipment)VAcuum systems
Standard Taper Joint34/35, 19/26
Unit SizeEA