1x1 image pixel for data collection
epTIPS™ Low-retention Pipettor Tips
epTIPS™ Low-retention Pipettor Tips
Actual product may vary
Fisherbrand
epTIPS™ Low-retention Pipettor Tips
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tips are ideal for cell culture (media) applications and applications with samples that contain detergents.
Have Questions?
Catalog NumberVolume (Metric)
FKS1053625250 to 1000 μL
Catalog number FKS10536252
Price (SGD)
202.00
PK
Add to cart
Volume (Metric):
50 to 1000 μL
Price (SGD)
202.00
PK
Add to cart
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tips are ideal for cell culture (media) applications and applications with samples that contain detergents.
  • Ultrahomogenous surface for maximum reproducibility
  • Ultrahydrophobic surface for minimal sample loss
  • Significantly reduced foam formation during pipetting
  • Extreme resistance to chemicals
  • Available in PCR clean and Eppendorf Quality
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free)
  • No coating
Specifications
Volume (Metric)50 to 1000 μL
FilteredNon-filtered
SterilitySterile
ColorBlue
Length (Metric)7.1cm
For Use With (Application)They satisfy the most stringent demands in medicine, the pharmaceutical and food industries, PCR*, molecular biology and cytotechnology
Tip StyleLow Retention
For Use With (Equipment)Eppendorf Pipetter
Product LineepT.I.P.S.
Unit SizePK
Share catalog number, name or link