Pipette tips, 500 - 5,000 μL, PP, Colorless, Certified Life Science Quality
Pipette tips, 500 - 5,000 μL, PP, Colorless, Certified Life Science Quality
Actual product may vary
Fisher Catalogue
Pipette tips, 500 - 5,000 μL, PP, Colorless, Certified Life Science Quality
Perfect fit for the Transferpette™ micropipette and many other pipettes. The tip system for all requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormat
FKS10530072Bulk
FKS10135350TipBox
FIS04-977-160Bulk
Catalog number FKS10530072
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Format:
Bulk
Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses. The raw materials are free from DiHEMDA and Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. BRAND offers pipette tips for volumes from 0.1 μl to 10 ml and filter tips for volumes from 0.1 μl to 1250 μl. They are tested for BRAND pipettes and many pipette from other manufacturers.
  • BRAND pipette tips and filter tips as well as the corresponding TipBoxes are autoclavable at 121 °C (2 bar) acc. DIN EN 285.
  • Sterile tips are manufactured under certified BIO-CERT™ quality. They are sterilized with beta radiation according to ISO 11 137 and the AAMI Guidelines.
  • Certified Life Science Quality: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
Specifications
Volume (Metric)0.5 to 5 mL
SterilityNon-sterile
ColorColorless
AutoclavableAutoclavable
Length (Metric)16 cm
Clean ClaimsDiHEMDA- and oleamide-free
FormatBulk
Tip StyleSlender
PackagingBagged, 200 Tips
For Use With (Equipment)Transferpette™, Thermo Scientific™ Finnpipette™ Microliter Pipette