Check Order Status
Fluted Interior PP Funnels
Actual product may vary
Fisherbrand
Fluted Interior PP Funnels
External flutings allow air to escape during filtration.
Have Questions?
Catalog Number
Diameter (Metric)Top
FKS10528331
100mm
Catalog number
FKS10528331
Price (SGD)
24.00
PK
Add to cart
Diameter (Metric)Top:
100mm
Price (SGD)
24.00
PK
Add to cart
Manufactured from polypropylene for good chemical resistance
For rapid filtration
Angled at 60°
Specifications
Diameter (Metric)Top
100mm
Diameter (Metric) Stem
11mm
Material
Polypropylene
Angle
60°
Color
Translucent
For Use With (Application)
Rapid filtration
Length (Metric) Stem
82mm
Quantity
1/Pk.
Product Type
Funnel