Features a PEI body, double junction pH electrode with a BNC connector that has a design consideration that is ideal for field applications and general purpose use.
Have Questions?
Catalog Number
Length (Metric)
FKS10527912
120 mm
Catalog number FKS10527912
Price (SGD)
649.00
EA
Add to cart
Length (Metric):
120 mm
Price (SGD)
649.00
EA
Add to cart
Is a PEI body, double junction pH electrode with a BNC connector.
Electrode features a single ceramic junction in the outer reference cell
Spherical pH sensing portion is made with general purpose glass and is protected by an extension of the durable PEI body
Design consideration is ideal for field applications as well as general purpose use
Uses general purpose (GP) glass, spherical bulb, polyetherimide (PEI) body, single ceramic frit, and double junction