Check Order Status
Stainless Steel Stand Base
Actual product may vary
Fisherbrand
Stainless Steel Stand Base
Support your supplies with the Bochem™ Stainless Steel Stand Base, which has a reinforcing leg for stability.
Have Questions?
Catalog Number
Length (Metric)
Width (Metric)
FKS10527561
315 mm
200 mm
Catalog number
FKS10527561
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
315 mm
Width (Metric):
200 mm
Expanded, flat and heavy feet for stability
1 adjustable foot
Anti-slide rubber protection
Thread M10
Specifications
Material
Stainless Steel (18/10)
Height (Metric)
3 mm
Length (Metric)
315 mm
Width (Metric)
200 mm
Product Type
Stand Base
Quantity
1 Pk.