1x1 image pixel for data collection
96-Well, Non-Treated, U-Shaped-Bottom Microplate
96-Well, Non-Treated, U-Shaped-Bottom Microplate
Actual product may vary
Fisherbrand
96-Well, Non-Treated, U-Shaped-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Have Questions?
Catalog NumberColor
FKS10520832Clear
Catalog number FKS10520832
Price (SGD)
709.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Color:
Clear
Price (SGD)
709.00
PK
Add to cart
Available with tissue culture-treated and nontreated surfaces. Made of crystal grade virgin polystyrene for optimum visibility.
  • Round bottom, with lid, individually wrapped, sterile
  • Crystal-grade virgin polystyrene
  • Sterilized by gamma irradiation
  • Nonpyrogenic
  • Non-treated polystyrene, standard tissue culture (TC), and Corning™ PrimariaTM surface treatments available
  • Convenient, peel-open medical-style packaging for individual product presentation
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
  • Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
Specifications
FormatStandard
LidWith Lid
Culture Area0.36 cm2
Array8 x 12
Binding TypeNot Treated
ColorClear
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working1.5 to 2.2 mL
SterilitySterile
Surface TreatmentNon-treated
Total Volume (Metric) Well0.32 mL
Well ShapeRound Bottom (U-well)
No. per Case50
Quantity50/Cs.
Unit SizePK
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link