Orion™ Ammonia Combination Ion Selective Electrode
Orion™ Ammonia Combination Ion Selective Electrode
Actual product may vary
Orion™ Ammonia Combination Ion Selective Electrode
Orion™ Ammonia Combination Ion Selective Electrode
Orion™ Ammonia Combination Ion Selective Electrode
Orion™ Ammonia Combination Ion Selective Electrode
Orion™ Ammonia Combination Ion Selective Electrode
Fisherbrand
Orion™ Ammonia Combination Ion Selective Electrode
Get quick, reliable and accurate measurements of ammonia.
Have Questions?
Catalog NumberConnector Types
FKS10518941BNC (Waterproof)
Catalog number FKS10518941
Price (SGD)
3,617.00
EA
Add to cart
Connector Types:
BNC (Waterproof)
Price (SGD)
3,617.00
EA
Add to cart
Get quick, reliable and accurate measurements of dissolved ammonia in aqueous solutions with the Thermo Scientific™ Orion™ Ammonia Gas Sensing ISE Electrode.
Ammonia is an important aspect of wastewater and environmental management. The 9512BNWP complies with SM 4500-NH3 D-1997 or E-1997; ASTM D1426-08 (B).
Features:
  • 9512BNWP features a hydrophobic gas-permeable membrane technology for use with either loose membrane 951204 or pre-assembled membrane caps 951205. Good for samples with > 0.1 mg/L ammonia.
Solutions required:
  • Standards
    • 951006 0.1 M NH4Cl standard, 475 mL bottle
    • 951007 1000 ppm ammonia as nitrogen standard, 475 mL bottle
    • 951207 100 ppm ammonia as nitrogen standard, 475 mL bottle
  • ISA
    • 951211 Ammonia ISA with blue dye, 475 mL bottle
  • Electrode fill solution
    • 951202 Electrode filling solution, 60 ml bottle
  • Storage solution
    • 951213 Ammonia electrode storage solution, 475 mL bottle
Consumables required:
  • Membranes
    • 951204 Loose membranes for standard electrodes, box of 20
    • 951205 Bonded membranes for standard electrodes, pack of 3
  • Items included with
    • 951202 Electrode filling solution, 60 mL 1 bottle
    • 951204 Loose membranes, box of 20. 2 boxes
Warranty period:
  • 12 months from date of purchase
Specifications
Connector TypesBNC (Waterproof)
DescriptionAmmonia Gas Sensing ISE Electrode
Length (English)5.87 in.
Length (English) Cable29.5 in.
Length (Metric)15.1cm
Length (Metric) Cable75cm
MaterialEpoxy
Operating Range (Metric)Ranges :1.0 to 5 x 10E7M
Temperature (English) Operating32° to 122°F
Temperature (Metric) Operating0°C to 50°C
TypeAmmonia Gas-Sensing ISE Electrode
Unit SizeEA