Short-Term Detector Tubes: Xylene
Short-Term Detector Tubes: Xylene
Actual product may vary
Fisherbrand
Short-Term Detector Tubes: Xylene
Detector tubes
Have Questions?
Catalog number FKS10518682
Price (SGD)
551.00
PK
Add to cart
Price (SGD)
551.00
PK
Add to cart
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Specifications
Components MeasuredXylene
Measurement Range10 to 400
No. of Strokes5
Quantity10
Response Time2.3 min.
Temperature Range (English)41°F to 95°F
Temperature Range (Metric)5° to 35°C
Tube TypeXylene 10/a
Unit SizePK