Digital Hand-Held Pocket Refractometer, PAL-1
Digital Hand-Held Pocket Refractometer, PAL-1
Actual product may vary
Digital Hand-Held Pocket Refractometer, PAL-1
Digital Hand-Held Pocket Refractometer, PAL-1
Fisherbrand
Digital Hand-Held Pocket Refractometer, PAL-1
Wide measurement range. The Atago Digital Hand-Held Pocket Refractometer, PAL-1 is ideal for measuring almost any fruit juice, food or drink.
Have Questions?
Catalog number FKS10510443
Price (SGD)
1,433.00
PK
Add to cart
Price (SGD)
1,433.00
PK
Add to cart
For measuring fruit juice, food, or drink, such as soup, sauce, ketchup, tomato sauce, low-sugar jam, or marmalade.
Specifications
Scale Range0.0 to 53.0 Brix
ModelPAL-1
Sample Volume0.3 mL
CalibrationWater
Accuracy±0.2% Brix
Repeatability±0.1% (Brix)
Resolution0.1% Brix
DisplayLCD
Scale TypeBrix
Measurement Time3 sec.
For Use With (Application)Measuring fruit juice, food, or drink, such as soup, sauce, ketchup, tomato sauce, low-sugar jam, or marmalade
IP RatingIP65 water resistant
Weight (English)0.22 lb.
Weight (Metric)100 g
Height (English)4.3 in.
Height (Metric)109 mm
Width (English)2.2 in.
Width (Metric)55 mm
Ambient Temperature (Metric)10°C to 40°C
Temperature Accuracy±1°C
Temperature Compensation10°C to 100°C
Temperature Range (Metric)10°C to 40°C
Temperature Resolution0.1°C
Power Supply Details2xAAA Batteries
Unit SizePK