Minisart™ NML Syringe Filters, Sterile
Minisart™ NML Syringe Filters, Sterile
Actual product may vary
Fisher Catalogue
Minisart™ NML Syringe Filters, Sterile
Provides fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units. Sartorius Minisart™ NML Syringe Filters, Sterile eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeColorSterilization MethodOutlet Style
FKS105098210.2 μmBlueETO-sterileMale Luer Lock
FIS14-555-2700.2 μmBlueETO-sterileMale Luer Lock
FIS14-555-2690.2 μmBlueGamma-sterile, individually packagedMale Luer Lock
FKS101091800.45 μmYellowETO-sterileMale Luer Lock
Catalog number FKS10509821
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.2 μm
Color:
Blue
Sterilization Method:
ETO-sterile
Outlet Style:
Male Luer Lock
Provides fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units. Sartorius Minisart™ NML Syringe Filters, Sterile eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system.
Fast and reliable filtration of small to medium volumes, complete with ready-to-connect filter units eliminate time consuming preparatory work, such as changing out the filter and cleaning the filtration system. These labor saving units are used for a wide variety of applications: sample cleaning, particle removal, and sterile filtration.
  • Handy and readily available
  • Easy to use
  • Proven quality
  • Economical
  • Available in a wide array of different types
  • 100% tested during manufacture
Specifications
ColorBlue
DisposableSingle-use
Filter TypeSyringe Filter
For Use With (Equipment)Syringe
Material (Membrane)SFCA
Pore Size0.2 μm
Sample Volume1 to 200 mL
SterilitySterile
Filtration Area6.2 cm2
For Use With (Application)Preparation of aqueous liquids
Hold up Volume (After Air Purge)0.15 mL
Housing MaterialMeta acrylate butadiene styrene polymerisate (MBS)
Inlet StyleFemale Luer Lock
Outlet StyleMale Luer Lock
Quantity50/Pk.
Sterilization MethodETO-sterile
Temperature Range (Metric)50°C
TypeNML Syringe Filter
Volume (Metric)200 mL
Dimensions (L x W x H)200 x 130 x 100 mm
Diameter (Metric)28 mm
Unit SizePK