Polypropylene Graduated Microtubes
Polypropylene Graduated Microtubes
Actual product may vary
Fisherbrand
Polypropylene Graduated Microtubes
Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. Eppendorf™ Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorCapacity (Metric)Purity or Quality Grade
FKS10509691Clear1.5 mLEppendorf Quality
AIT0030121155Amber0.5 mLPCR Clean
FKS10458312Amber0.5 mLPCR Clean
FKS10252851Amber2 mLPCR Clean
FKS10241931Clear2 mLBiopur
AIT0030121023Clear0.5 mLEppendorf Quality
FKS10292611Clear0.5 mLEppendorf Quality
AIT0030120094Clear2 mLEppendorf Quality
FKS10038760Clear2 mLEppendorf Quality
AIT0030120086Clear1.5 mLEppendorf Quality
Catalog number FKS10509691
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Clear
Capacity (Metric):
1.5 mL
Purity or Quality Grade:
Eppendorf Quality
Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. EppendorfTM Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
  • Biohazardous substances can be removed, free of aerosols, through a thin membrane in the center of the lid with a cannula
  • Lid easily opens with one hand
  • These tubes for 0.5mL, 1.5mL and 2mL have a graduation scale and writing surfaces
  • The 0.5mL tube is optimized for use in PCR* experiments
  • Assorted color packs are a mixture of yellow, red, blue, green and colorless
  • Autoclavable micropestle for re-suspending pellets
  • Can be centrifuged at up to 23,500xgr
Specifications
AutoclavableAutoclavable
Capacity (Metric)1.5 mL
ColorClear
MaterialPolypropylene
For Use With (Application)Applications requiring high general quality, but no special purities
For Use With (Equipment)Centrifugation
No. per Pack1000/Pk.
Purity or Quality GradeEppendorf Quality
GraduatedYes
Closure TypeHinged Snap Cap
Max. RCF30,000 x g
Unit SizePK