Nunc™ Cell-Culture Treated Multidishes
Nunc™ Cell-Culture Treated Multidishes
Actual product may vary
Fisher Catalogue
Nunc™ Cell-Culture Treated Multidishes
Reduce the risk of contamination with these multidishes, which feature a raised well rim and a special surface treatment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsNo. per Case
FKS105075914120
FKS10119831675
FIS12-565-1632475
FKS100988701275
FIS12-565-724120
FKS10469282685
FIS12-565-1642485
FKS103769122485
Catalog number FKS10507591
View Price:Sign in to see your account pricing. Need an account? Register with us today.
No. of Wells:
4
No. per Case:
120
Promote maximum adhesion for a broad range of cell types with the Thermo Scientific™ Nunc™ Cell-Culture Treated multi-well plates with the Nunclon™ Delta surface treatment. The open format with the raised well rim enables convenient and secure manipulation of the cells.
Features:
  • Flat well bottom and stackable
  • Raised well rims to lower the risk of cross contamination
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Excellent optical quality
  • Alphanumerical codes for well identification
Round Well Styles:
  • Feature vented lids for gas exchange
  • Recessed areas around wells act as a water reservoir to reduce evaporation from wells
  • Lids with notched corners for easy orientation
Rectangular Well Styles:
  • Large culture area
  • Fit standard glass cover slips: 24×60mm in 4-well plate; and 22×22mm in 8-well plate
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceISO 13485:2016 (BSI Certificate Number: FM 653694)
FormatStandard
Culture Area1.9 sq. cm
Array2 x 2
ColorClear
Product TypeCell Culture Dish
MaterialPolystyrene
Volume (Metric) Well1 mL (Well)
No. of Wells4
Volume (Metric) Working1mL
SterilitySterile
Surface TreatmentNunclon Delta
Well ShapeFlat Bottom (F-well)
IncludesLid
No. per Case120
No. per Pack4
Unit SizePK