Polypropylene Stirring Rods with Flattened Ends
Polypropylene Stirring Rods with Flattened Ends
Actual product may vary
Fisherbrand
Polypropylene Stirring Rods with Flattened Ends
Manufactured from polypropylene for strength and durability. Kartell™ Polypropylene Stirring Rods with Flattened Ends provides two distinct shapes for stirring and scraping applications.
Have Questions?
Catalog NumberLength (Metric)
FKS10505241245 mm
Catalog number FKS10505241
Price (SGD)
32.00
PK
Add to cart
Length (Metric):
245 mm
Price (SGD)
32.00
PK
Add to cart
Manufactured from polypropylene for strength and durability. Kartell™ Polypropylene Stirring Rods with Flattened Ends provides two distinct shapes for stirring and scraping applications.
  • Tapered body gives strength to the 12mm triangular scraper at one end and flexibility to a 15mm palette knife blade at the other
  • White color
Specifications
Length (Metric)245 mm
MaterialPolypropylene
TypeStirring Rod
Unit SizePK