DURAN™ Blanking Screw Cap, GL 14, Red PBT, with PTFE Cap Liner (GL 45 Connection Cap System Accessory)
DURAN™ Blanking Screw Cap, GL 14, Red PBT, with PTFE Cap Liner (GL 45 Connection Cap System Accessory)
Actual product may vary
Fisher Catalogue
DURAN™ Blanking Screw Cap, GL 14, Red PBT, with PTFE Cap Liner (GL 45 Connection Cap System Accessory)
Accessory for 2 or 3 port GL 45 connection cap system
Have Questions?
Catalog number FKS10501125
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Flexible modular GL 45 connection cap system that accepts four different rigid or semi-rigid tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm). The caps are available with 2 or 3 GL 14 threaded ports. The different sized inserts form a tight seal around the pass-through tubing. Sterile pressure equalisation is possible through use of 0.2 micron syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.
  • Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts
  • Available with 2, or 3 threaded outlets (ports)
  • Five different tubing diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be used by changing the inserts
  • By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap
  • Fully compatible with all the GL 45 threads and therefore highly versatile
  • Fully compatible with all the GLS 80 threads and therefore highly versatile
Suitable Accessories:
  • 1129750
  • 1129751
Specifications
DescriptionScrew cap,PBT, with PTFE cap liner, red, GL 14
For Use With (Application)Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply
MaterialPolypropylene, PTFE
Quantity2/Pk.
Thread Size (Metric)GL 14
AutoclavableAutoclavable
Diameter (Metric)87 mm
Unit SizePK